Assay Details
Target Gene Details
Entrez Gene ID: | 27068 |
Gene Name: | pyrophosphatase (inorganic) 2 |
Gene Aliases: |
HSPC124, SID6-306 |
Location: |
Chr.4:105369077-105474070 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPA2 | NM_006903.4 | NP_008834.3 | ||
| NM_176866.2 | NP_789842.2 | |||
| NM_176867.3 | NP_789843.2 | |||
| NM_176869.2 | NP_789845.1 | |||
| AB026722.1 | BAA84701.1 | |||
| AF161473.1 | AAF29088.1 | |||
| AF217187.1 | AAG36781.1 | |||
| AK000466.1 | BAA91184.1 | |||
| AK289607.1 | ||||
| AL136655.1 | CAB66590.2 | |||
| BC022803.1 | AAH22803.1 | |||
| BC039462.1 | AAH39462.2 | |||
| BC057219.1 | AAH57219.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1000196 | Chr.4:103475625 - 106148054 on Build GRCh38 | Loss |
|
| nsv470063 | Chr.4:104948391 - 105445052 on Build GRCh38 | Loss |
|
| nsv830027 | Chr.4:105300853 - 105485581 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs66763438] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map