Assay Details
Target Gene Details
Entrez Gene ID: | 291 |
Gene Name: | solute carrier family 25 member 4 |
Gene Aliases: |
AAC1, ANT, ANT 1, ANT1, MTDPS12, PEO2, PEO3, PEOA2, T1 |
Location: |
Chr.4:185143263-185150384 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A4 | NM_001151.3 | 4 | 3456 | NP_001142.2 |
| AF052119.1 | 1 | 1420 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv461859 | Chr.4:184907523 - 185314394 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map