Assay Details
Target Gene Details
Entrez Gene ID: | 256471 |
Gene Name: | major facilitator superfamily domain containing 8 |
Gene Aliases: |
CCMD, CLN7 |
Location: |
Chr.4:127917805-127965984 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFSD8 | NM_152778.2 | NP_689991.1 | ||
| XM_005262893.1 | XP_005262950.1 | |||
| XM_005262896.1 | XP_005262953.1 | |||
| XM_005262897.2 | XP_005262954.1 | |||
| XM_005262898.2 | XP_005262955.1 | |||
| XM_011531830.1 | XP_011530132.1 | |||
| XM_011531831.1 | XP_011530133.1 | |||
| XM_011531832.1 | XP_011530134.1 | |||
| XM_017007989.1 | XP_016863478.1 | |||
| AK074564.1 | BAC11062.1 | |||
| AK294184.1 | ||||
| AK294439.1 | ||||
| AK294527.1 | ||||
| AK294957.1 | ||||
| AK315596.1 | ||||
| BC029503.1 | AAH29503.1 | |||
| BI553701.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5385n100 | Chr.4:127882004 - 128173863 on Build GRCh38 | Gain |
|
| nsv1019193 | Chr.4:127889372 - 128028165 on Build GRCh38 | Loss |
|
| nsv595388 | Chr.4:127932758 - 128163676 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map