Assay Details
Target Gene Details
Entrez Gene ID: | 84068 |
Gene Name: | solute carrier family 10 member 7 |
Gene Aliases: |
C4orf13, P7 |
Location: |
Chr.4:146253981-146522351 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC10A7 | NM_001029998.5 | NP_001025169.1 | ||
| NM_001300842.2 | NP_001287771.1 | |||
| NM_001317816.1 | NP_001304745.1 | |||
| NR_133924.1 | ||||
| XM_011532311.2 | XP_011530613.1 | |||
| XM_011532313.2 | XP_011530615.1 | |||
| XM_011532314.2 | XP_011530616.1 | |||
| AK075364.1 | BAC11571.1 | |||
| AK126000.1 | ||||
| AK172727.1 | ||||
| AK291046.1 | ||||
| AY346324.1 | AAQ84722.1 | |||
| BC150308.1 | ||||
| CR933647.1 | CAI45948.1 | |||
| CR936751.1 | ||||
| DQ122860.1 | AAZ32256.1 | |||
| DQ122861.1 | AAZ32257.1 | |||
| DQ871036.1 | ABI31650.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv595651 | Chr.4:146195712 - 146847054 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map