Assay Details
Target Gene Details
Entrez Gene ID: | 83894 |
Gene Name: | tetratricopeptide repeat domain 29 |
Gene Aliases: |
NYD-SP14, TBPP2A |
Location: |
Chr.4:146706638-146945886 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC29 | NM_001300761.2 | NP_001287690.1 | ||
| NM_001317806.1 | NP_001304735.1 | |||
| NM_031956.3 | NP_114162.2 | |||
| NR_133922.1 | ||||
| XM_005263270.1 | XP_005263327.1 | |||
| XM_006714335.1 | XP_006714398.1 | |||
| XM_006714336.1 | XP_006714399.1 | |||
| XM_006714339.2 | XP_006714402.1 | |||
| AF345910.1 | AAK29064.1 | |||
| AK093145.1 | BAC04072.1 | |||
| AK225725.1 | ||||
| AK301807.1 | ||||
| AK314066.1 | ||||
| BC024193.2 | AAH24193.1 | |||
| EF432564.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3602584 | Chr.4:146683951 - 146768781 on Build GRCh38 | Loss |
|
| nsv595651 | Chr.4:146195712 - 146847054 on Build GRCh38 | Gain |
|

Genomic Map