Assay Details
Target Gene Details
Entrez Gene ID: | 51091 |
Gene Name: | Sep (O-phosphoserine) tRNA:Sec (selenocysteine) tRNA synthase |
Gene Aliases: |
LP, PCH2D, SLA, SLA/LP |
Location: |
Chr.4:25120005-25160582 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEPSECS | NM_016955.3 | NP_058651.3 | ||
| XM_011513846.2 | XP_011512148.1 | |||
| XM_011513847.2 | XP_011512149.1 | |||
| XM_011513848.1 | XP_011512150.1 | |||
| XM_017008277.1 | XP_016863766.1 | |||
| XM_017008278.1 | XP_016863767.1 | |||
| AF146396.2 | AAD33963.2 | |||
| AF282065.1 | AAG00491.1 | |||
| AJ238617.1 | CAB62209.1 | |||
| AJ277541.1 | CAB89517.1 | |||
| AK292476.1 | ||||
| BC117202.1 | ||||
| BC126213.1 | ||||
| BX648976.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4277 | Chr.4:25127698 - 25172730 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77856916] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map