Assay Details
Target Gene Details
Entrez Gene ID: | 3660 |
Gene Name: | interferon regulatory factor 2 |
Gene Aliases: |
IRF-2 |
Location: |
Chr.4:184387722-184474572 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IRF2 | NM_002199.3 | NP_002190.2 | ||
| AK312953.1 | ||||
| BC015803.2 | AAH15803.1 | |||
| BT007264.1 | AAP35928.1 | |||
| BX648934.1 | 1 | 172 | ||
| CR457077.1 | CAG33358.1 | |||
| X15949.1 | CAA34073.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1026074 | Chr.4:184384963 - 184780663 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map