Assay Details
Target Gene Details
Entrez Gene ID: | 57620 |
Gene Name: | stromal interaction molecule 2 |
Gene Aliases: |
- |
Location: |
Chr.4:26860691-27025381 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STIM2 | NM_001169117.1 | NP_001162588.1 | ||
| NM_001169118.1 | NP_001162589.1 | |||
| NM_020860.3 | NP_065911.3 | |||
| AB040915.1 | ||||
| AF328905.1 | AAK82337.1 | |||
| AK096846.1 | ||||
| AL136577.1 | CAB66512.2 | |||
| BC136449.1 | ||||
| BC146661.1 | ||||
| BC152554.1 | ||||
| BC171766.1 | ||||
| BX343313.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv461312 | Chr.4:26843052 - 27076754 on Build GRCh38 | Loss |
|
| nsv470021 | Chr.4:26719956 - 27065405 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map