Assay Details
Target Gene Details
Entrez Gene ID: | 643749 |
Gene Name: | TRAF3IP2 antisense RNA 1 |
Gene Aliases: |
C6UAS, C6orf3, NCRNA00248, TRAF3IP2-AS2 |
Location: |
Chr.6:111483472-111602295 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAF3IP2-AS1 | NR_034108.1 | |||
| NR_034109.1 | ||||
| NR_034110.1 | ||||
| NR_034111.1 | ||||
| BX647114.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs144014529] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map