Assay Details
Target Gene Details
Entrez Gene ID: | 57150 |
Gene Name: | small integral membrane protein 8 |
Gene Aliases: |
C6orf162, dJ102H19.2 |
Location: |
Chr.6:87322588-87342328 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMIM8 | NM_001042493.2 | NP_001035958.1 | ||
| NM_001287445.1 | NP_001274374.1 | |||
| NM_020425.5 | NP_065158.3 | |||
| AF311339.1 | AAK38513.1 | |||
| AK311963.1 | ||||
| BC070260.1 | AAH70260.1 | |||
| BG534658.1 | ||||
| DA293899.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5386 | Chr.6:87328833 - 87335203 on Build GRCh38 | Deletion |
|

Genomic Map