Assay Details
Target Gene Details
Entrez Gene ID: | 26575 |
Gene Name: | regulator of G-protein signaling 17 |
Gene Aliases: |
RGS-17, RGSZ2, hRGS17 |
Location: |
Chr.6:153010891-153131284 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGS17 | NM_012419.4 | NP_036551.3 | ||
| XM_017010733.1 | XP_016866222.1 | |||
| AF202257.3 | AAF08978.3 | |||
| BC013117.1 | AAH13117.1 | |||
| BT006997.1 | AAP35643.1 |
More Information
Set Membership: |
|

Genomic Map