Assay Details
Target Gene Details
Entrez Gene ID: | 100874531 |
Gene Name: | long intergenic non-protein coding RNA 581 |
Gene Aliases: |
linc-MBOAT1-4 |
Location: |
Chr.6:21486061-21511892 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00581 | NR_103790.1 | 4 | 657 | |
| AA910749.1 | 1 | 339 | ||
| BX118339.1 | 4 | 661 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830601 | Chr.6:21321631 - 21509567 on Build GRCh38 | Loss |
|
| esv2762582 | Chr.6:21474503 - 21506299 on Build GRCh38 | Loss |
|

Genomic Map