Assay Details
Target Gene Details
Entrez Gene ID: | 2822 |
Gene Name: | glycosylphosphatidylinositol specific phospholipase D1 |
Gene Aliases: |
GPIPLD, GPIPLDM, PIGPLD, PIGPLD1, PLD |
Location: |
Chr.6:24425832-24495163 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPLD1 | NM_001503.3 | 25 | 4498 | NP_001494.2 |
| XM_017010753.1 | 26 | 4570 | XP_016866242.1 | |
| XM_017010754.1 | 20 | 3989 | XP_016866243.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1026638 | Chr.6:24347744 - 24475072 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map