Assay Details
Target Gene Details
Entrez Gene ID: | 81491 |
Gene Name: | G protein-coupled receptor 63 |
Gene Aliases: |
PSP24(beta), PSP24B |
Location: |
Chr.6:96794122-96837477 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR63 | NM_001143957.2 | 3 | 2521 | NP_001137429.1 |
| NM_030784.3 | 2 | 2403 | NP_110411.1 | |
| XM_006715570.3 | 2 | 2473 | XP_006715633.1 | |
| XM_011536153.2 | 3 | 2464 | XP_011534455.1 | |
| XM_011536154.2 | 3 | 2337 | XP_011534456.1 | |
| XM_011536155.2 | 3 | 2516 | XP_011534457.1 | |
| XM_011536157.2 | 2 | 2239 | XP_011534459.1 | |
| XM_011536159.2 | 2 | 2408 | XP_011534461.1 | |
| XM_017011334.1 | 2 | 2931 | XP_016866823.1 |
More Information
Set Membership: |
|

Genomic Map