Assay Details
Target Gene Details
Entrez Gene ID: | 6196 |
Gene Name: | ribosomal protein S6 kinase A2 |
Gene Aliases: |
HU-2, MAPKAPK1C, RSK, RSK3, S6K-alpha, S6K-alpha2, p90-RSK3, p90RSK2, pp90RSK3 |
Location: |
Chr.6:166409364-166862551 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS6KA2 | NM_001006932.2 | NP_001006933.2 | ||
| NM_001318936.1 | NP_001305865.1 | |||
| NM_001318937.1 | NP_001305866.1 | |||
| NM_001318938.1 | NP_001305867.1 | |||
| NM_021135.5 | NP_066958.2 | |||
| XM_006715549.3 | XP_006715612.1 | |||
| AB209116.1 | BAD92353.1 | |||
| AK095751.1 | ||||
| AK295674.1 | ||||
| AK310428.1 | ||||
| BC002363.2 | AAH02363.1 | |||
| KJ534936.1 | ||||
| KJ535080.1 | ||||
| KJ535087.1 | ||||
| L07598.1 | AAC82496.1 | |||
| X85106.1 | CAA59427.1 |
Target Gene Details
Entrez Gene ID: | 100874353 |
Gene Name: | RPS6KA2 intronic transcript 1 |
Gene Aliases: |
- |
Location: |
Chr.6:166460663-166465383 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS6KA2-IT1 | NR_046793.1 | |||
| AW992696.1 |
More Information
Set Membership: |
|

Genomic Map