Assay Details
Target Gene Details
Entrez Gene ID: | 154089 |
Gene Name: | long intergenic non-protein coding RNA 1312 |
Gene Aliases: |
- |
Location: |
Chr.6:133821147-133853992 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01312 | NR_027030.1 |
Target Gene Details
Entrez Gene ID: | 100507308 |
Gene Name: | TCF21 antisense RNA inducing promoter demethylation |
Gene Aliases: |
EYA4-AS1 |
Location: |
Chr.6:133502252-133889006 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TARID | NR_109982.1 | |||
| DB063930.1 | ||||
| DB279014.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34122 | Chr.6:133767881 - 134057114 on Build GRCh38 | Loss |
|
| nsv830812 | Chr.6:133672759 - 133844269 on Build GRCh38 | Gain |
|
| esv2732724 | Chr.6:133832985 - 133833781 on Build GRCh38 | Deletion |
|
| esv2011634 | Chr.6:133833223 - 133833750 on Build GRCh38 | Deletion |
|

Genomic Map