Assay Details
Target Gene Details
Entrez Gene ID: | 9994 |
Gene Name: | caspase 8 associated protein 2 |
Gene Aliases: |
CED-4, FLASH, RIP25 |
Location: |
Chr.6:89829900-89874436 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASP8AP2 | NM_001137667.1 | NP_001131139.1 | ||
| NM_001137668.1 | NP_001131140.1 | |||
| NM_012115.3 | NP_036247.1 | |||
| AB037736.2 | BAA92553.2 | |||
| AF154415.1 | AAF03367.1 | |||
| AF164678.2 | AAD45537.2 | |||
| BC056685.1 | AAH56685.1 | |||
| BC132828.1 | ||||
| BC132830.1 | ||||
| BC150274.1 | ||||
| BP192859.1 | ||||
| BP363682.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5391 | Chr.6:89837037 - 89869200 on Build GRCh38 | Insertion |
|
| nsv604125 | Chr.6:89751706 - 90189842 on Build GRCh38 | Gain |
|
| esv3890901 | Chr.6:89204468 - 90033564 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs80210942] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map