Assay Details
Target Gene Details
Entrez Gene ID: | 9263 |
Gene Name: | serine/threonine kinase 17a |
Gene Aliases: |
DRAK1 |
Location: |
Chr.7:43583093-43627379 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK17A | NM_004760.2 | NP_004751.2 | ||
| XM_017012792.1 | XP_016868281.1 | |||
| AB011420.1 | BAA34126.1 | |||
| BC023508.2 | AAH23508.2 | |||
| BC047696.1 | AAH47696.1 | |||
| BI461892.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3612993 | Chr.7:43438543 - 43609526 on Build GRCh38 | Gain |
|
| nsv518728 | Chr.7:43555801 - 43614160 on Build GRCh38 | Loss |
|
| esv3612996 | Chr.7:43537785 - 43656356 on Build GRCh38 | Gain |
|
| dgv11293n54 | Chr.7:43583370 - 43604236 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map