Assay Details
Target Gene Details
Entrez Gene ID: | 93986 |
Gene Name: | forkhead box P2 |
Gene Aliases: |
CAGH44, SPCH1, TNRC10 |
Location: |
Chr.7:114086310-114693772 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXP2 | NR_033766.1 | |||
| XM_017012801.1 | XP_016868290.1 | |||
| AF454830.1 | AAM60761.1 | |||
| AK131266.1 | BAD18444.1 | |||
| CR749236.1 | CAH18092.1 | |||
| DA400243.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv608231 | Chr.7:114067732 - 114419101 on Build GRCh38 | Loss |
|
| nsv608230 | Chr.7:113941914 - 114186383 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map