Assay Details
Target Gene Details
Entrez Gene ID: | 7791 |
Gene Name: | zyxin |
Gene Aliases: |
ESP-2, HED-2 |
Location: |
Chr.7:143381174-143391111 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZYX | NM_001010972.1 | NP_001010972.1 | ||
| NM_003461.4 | NP_003452.1 | |||
| XM_011516569.2 | XP_011514871.2 | |||
| XM_017012587.1 | XP_016868076.1 | |||
| AK130138.1 | ||||
| AK298925.1 | ||||
| AK299005.1 | ||||
| AK302309.1 | ||||
| AK316227.1 | ||||
| BC002323.2 | AAH02323.2 | |||
| BC008743.2 | AAH08743.1 | |||
| BC009360.2 | AAH09360.1 | |||
| BC010031.2 | AAH10031.1 | |||
| BC017183.2 | AAH17183.2 | |||
| CR457431.1 | CAG33712.1 | |||
| U15158.1 | AAA78902.1 | |||
| X94991.1 | CAA64447.1 | |||
| X95735.1 | CAA65050.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv949967 | Chr.7:143382508 - 143399907 on Build GRCh38 | Deletion |
|
| nsv482115 | Chr.7:143381267 - 143391109 on Build GRCh38 | Gain |
|
| nsv608681 | Chr.7:143372217 - 143425364 on Build GRCh38 | Loss |
|
| nsv517520 | Chr.7:143352880 - 143400007 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map