Assay Details
Target Gene Details
Entrez Gene ID: | 11112 |
Gene Name: | 3-hydroxyisobutyrate dehydrogenase |
Gene Aliases: |
NS5ATP1 |
Location: |
Chr.7:27525440-27663001 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIBADH | NM_152740.3 | NP_689953.1 | ||
| XM_017011719.1 | XP_016867208.1 | |||
| AB050000.1 | ||||
| AF529362.1 | AAQ09596.1 | |||
| AK316605.1 | ||||
| BC032324.1 | AAH32324.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830930 | Chr.7:27356219 - 27547736 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map