Assay Details
Target Gene Details
Entrez Gene ID: | 94239 |
Gene Name: | H2A histone family member V |
Gene Aliases: |
H2A.Z-2, H2AV |
Location: |
Chr.7:44826889-44848126 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H2AFV | NM_138635.3 | NP_619541.1 | ||
| AK023973.1 | ||||
| BM452844.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830985 | Chr.7:44793475 - 44979588 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map