Assay Details
Target Gene Details
Entrez Gene ID: | 105375131 |
Gene Name: | basic proline-rich protein-like |
Gene Aliases: |
- |
Location: |
Chr.7:4126653-4137206 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105375131 | XM_011515667.2 | XP_011513969.1 |
Target Gene Details
Entrez Gene ID: | 221935 |
Gene Name: | sidekick cell adhesion molecule 1 |
Gene Aliases: |
- |
Location: |
Chr.7:3301448-4269000 on Build GRCh38 |
Assay Gene Location: | Within Intron 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDK1 | NM_001079653.1 | NP_001073121.1 | ||
| NM_152744.3 | NP_689957.3 | |||
| XM_011515188.1 | XP_011513490.1 | |||
| XM_011515189.1 | XP_011513491.1 | |||
| XM_011515190.1 | XP_011513492.1 | |||
| XM_017011837.1 | XP_016867326.1 | |||
| XM_017011838.1 | XP_016867327.1 | |||
| XM_017011839.1 | XP_016867328.1 | |||
| XM_017011840.1 | XP_016867329.1 | |||
| AK074083.1 | BAB84909.1 | |||
| AL833354.1 | ||||
| AY310398.1 | AAP75619.1 | |||
| DB065853.1 | ||||
| DB454012.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1019305 | Chr.7:3952014 - 4236372 on Build GRCh38 | Gain |
|
| nsv1018462 | Chr.7:4058044 - 4133981 on Build GRCh38 | Loss |
|
| nsv518299 | Chr.7:4077033 - 4246965 on Build GRCh38 | Loss |
|
| esv3611906 | Chr.7:4042548 - 4170582 on Build GRCh38 | Gain |
|
| esv2764033 | Chr.7:4081946 - 4132259 on Build GRCh38 | Gain |
|
| nsv950795 | Chr.7:4127069 - 4179668 on Build GRCh38 | Deletion |
|
| dgv797n27 | Chr.7:3915884 - 4573976 on Build GRCh38 | Gain |
|
| nsv527624 | Chr.7:3987904 - 4384423 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map