Assay Details
Target Gene Details
Entrez Gene ID: | 1021 |
Gene Name: | cyclin dependent kinase 6 |
Gene Aliases: |
MCPH12, PLSTIRE |
Location: |
Chr.7:92604921-92836627 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK6 | NM_001145306.1 | NP_001138778.1 | ||
| NM_001259.6 | NP_001250.1 | |||
| XM_006715835.2 | XP_006715898.1 | |||
| AK313491.1 | ||||
| BC052264.1 | AAH52264.1 | |||
| X66365.1 | CAA47008.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516592 | Chr.7:92635679 - 92650049 on Build GRCh38 | Gain+Loss |
|
| nsv831062 | Chr.7:92611920 - 92831297 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map