Assay Details
Target Gene Details
Entrez Gene ID: | 94239 |
Gene Name: | H2A histone family member V |
Gene Aliases: |
H2A.Z-2, H2AV |
Location: |
Chr.7:44826889-44848126 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H2AFV | NM_012412.4 | NP_036544.1 | ||
| NM_138635.3 | NP_619541.1 | |||
| NM_201436.2 | NP_958844.1 | |||
| NM_201516.2 | NP_958924.1 | |||
| NM_201517.2 | NP_958925.1 | |||
| AB209001.1 | BAD92238.1 | |||
| AF081192.1 | AAC31938.1 | |||
| AK023973.1 | ||||
| AK129989.1 | ||||
| AL520785.1 | ||||
| AL520811.3 | ||||
| BC000098.2 | AAH00098.1 | |||
| BC004274.1 | ||||
| BC014885.2 | AAH14885.1 | |||
| BC070169.1 | AAH70169.1 | |||
| BF242439.1 | ||||
| BG754026.1 | ||||
| BG773256.1 | ||||
| BI257891.1 | ||||
| BM452844.1 | ||||
| BM453663.1 | ||||
| CB148943.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830985 | Chr.7:44793475 - 44979588 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map