Assay Details
Target Gene Details
Entrez Gene ID: | 2918 |
Gene Name: | glutamate metabotropic receptor 8 |
Gene Aliases: |
GLUR8, GPRC1H, MGLUR8, mGlu8 |
Location: |
Chr.7:126438598-127253294 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3614931 | Chr.7:126334896 - 126534697 on Build GRCh38 | Loss |
|
| esv3891200 | Chr.7:124990785 - 126688176 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76711555] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map