Assay Details
Target Gene Details
Entrez Gene ID: | 2969 |
Gene Name: | general transcription factor IIi |
Gene Aliases: |
BAP135, BTKAP1, DIWS, GTFII-I, IB291, SPIN, TFII-I, WBS, WBSCR6 |
Location: |
Chr.7:74657665-74760692 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTF2I | NM_001163636.2 | NP_001157108.1 | ||
| NM_001518.4 | NP_001509.3 | |||
| NM_032999.3 | NP_127492.1 | |||
| NM_033000.3 | NP_127493.1 | |||
| NM_033001.3 | NP_127494.1 | |||
| AF015553.1 | AAB70791.1 | |||
| AF035737.1 | AAC08312.1 | |||
| AF038967.1 | AAC08313.1 | |||
| AF038968.1 | AAC08314.1 | |||
| AF038969.1 | AAC08315.1 | |||
| AK292832.1 | ||||
| AK294935.1 | ||||
| BC070484.1 | AAH70484.1 | |||
| BC099907.1 | AAH99907.1 | |||
| KJ534859.1 | ||||
| KJ534860.1 | ||||
| KJ534861.1 | ||||
| KJ534862.1 | ||||
| KJ534863.1 | ||||
| KJ534965.1 | ||||
| U77948.1 | AAB48826.1 | |||
| Y14946.1 | CAA75163.1 |
Target Gene Details
Entrez Gene ID: | 101926943 |
Gene Name: | uncharacterized LOC101926943 |
Gene Aliases: |
- |
Location: |
Chr.7:74688937-74728918 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101926943 | NR_110044.1 | |||
| DA836308.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517573 | Chr.7:74706683 - 74725717 on Build GRCh38 | Loss |
|
| nsv1032137 | Chr.7:74697471 - 75115763 on Build GRCh38 | Gain |
|
| nsv5794 | Chr.7:74713348 - 74748765 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map