Assay Details
Target Gene Details
Entrez Gene ID: | 84792 |
Gene Name: | family with sequence similarity 220 member A |
Gene Aliases: |
ACPIN1, C7orf70, SIPAR |
Location: |
Chr.7:6329409-6348959 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM220A | NM_001037163.1 | NP_001032240.1 | ||
| AF533707.1 | AAQ09020.1 | |||
| AK093156.1 | BAC04076.1 | |||
| AK311878.1 | ||||
| BC006110.1 | ||||
| BC140712.1 | ||||
| BC142977.1 | ||||
| BP354450.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1020236 | Chr.7:6209299 - 6650746 on Build GRCh38 | Gain |
|
| nsv436904 | Chr.7:5035574 - 6713282 on Build GRCh38 | Insertion |
|
| nsv1025381 | Chr.7:6317073 - 6370187 on Build GRCh38 | Loss |
|
| nsv1020251 | Chr.7:6269897 - 6340310 on Build GRCh38 | Loss |
|

Genomic Map