Assay Details
Target Gene Details
Entrez Gene ID: | 222256 |
Gene Name: | cadherin related family member 3 |
Gene Aliases: |
CDH28 |
Location: |
Chr.7:105876773-106036431 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDHR3 | XM_005250224.4 | XP_005250281.1 | ||
| XM_017011863.1 | XP_016867352.1 | |||
| XM_017011865.1 | XP_016867354.1 | |||
| XM_017011866.1 | XP_016867355.1 | |||
| XM_017011867.1 | XP_016867356.1 | |||
| AK125261.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1027213 | Chr.7:105798164 - 106382423 on Build GRCh38 | Gain |
|
| nsv608070 | Chr.7:105863755 - 105893289 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map