Assay Details
Target Gene Details
Entrez Gene ID: | 441191 |
Gene Name: | ring finger protein 216 pseudogene 1 |
Gene Aliases: |
RNF216L, hCG2040556 |
Location: |
Chr.7:4973985-4998169 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF216P1 | NR_015449.1 | |||
| NR_023384.1 | ||||
| NR_023385.1 | ||||
| AK303831.1 | ||||
| AK308526.1 | ||||
| AK309798.1 | ||||
| AK311517.1 | ||||
| BX538059.1 | ||||
| DC357738.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525289 | Chr.7:4963742 - 5044110 on Build GRCh38 | Loss |
|
| dgv6238n100 | Chr.7:4810816 - 5030133 on Build GRCh38 | Loss |
|
| nsv1019120 | Chr.7:4890022 - 5098778 on Build GRCh38 | Loss |
|
| nsv8033 | Chr.7:4762656 - 5046966 on Build GRCh38 | Loss |
|

Genomic Map