Assay Details
Target Gene Details
Entrez Gene ID: | 101927550 |
Gene Name: | uncharacterized LOC101927550 |
Gene Aliases: |
- |
Location: |
Chr.7:99013165-99036492 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927550 | NR_110102.1 | |||
| AK001533.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3614265 | Chr.7:98999684 - 99016092 on Build GRCh38 | Loss |
|
| nsv819767 | Chr.7:99014290 - 99014969 on Build GRCh38 | Gain |
|

Genomic Map