Assay Details
Target Gene Details
Entrez Gene ID: | 57016 |
Gene Name: | aldo-keto reductase family 1 member B10 |
Gene Aliases: |
AKR1B11, AKR1B12, ALDRLn, ARL-1, ARL1, HIS, HSI |
Location: |
Chr.7:134527592-134541414 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKR1B10 | NM_020299.4 | NP_064695.3 | ||
| XM_011516416.1 | XP_011514718.1 | |||
| XM_011516417.2 | XP_011514719.1 | |||
| AF052577.1 | AAC36465.1 | |||
| AF524864.1 | AAO13380.1 | |||
| BC008837.2 | AAH08837.1 | |||
| BT006794.1 | AAP35440.1 | |||
| CR541801.1 | CAG46600.1 | |||
| U37100.1 | AAC17469.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2669756 | Chr.7:134525343 - 134560371 on Build GRCh38 | Deletion |
|
| nsv1026859 | Chr.7:134512710 - 134558688 on Build GRCh38 | Loss |
|
| nsv1031083 | Chr.7:134516469 - 134620674 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map