Assay Details
Target Gene Details
Entrez Gene ID: | 5218 |
Gene Name: | cyclin dependent kinase 14 |
Gene Aliases: |
PFTAIRE1, PFTK1 |
Location: |
Chr.7:90595994-91210590 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK14 | NM_001287135.1 | NP_001274064.1 | ||
| XM_005250436.3 | XP_005250493.1 | |||
| XM_011516306.2 | XP_011514608.1 | |||
| XM_011516307.2 | XP_011514609.1 | |||
| XM_017012320.1 | XP_016867809.1 | |||
| AF119833.1 | AAG43234.1 | |||
| AK297974.1 | ||||
| DC420282.1 | ||||
| DQ655962.2 | ||||
| DQ655963.2 | ||||
| DQ655965.2 | ||||
| DQ655966.2 | ||||
| DQ656060.2 | ||||
| HY179664.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891163 | Chr.7:90335679 - 90681223 on Build GRCh38 | Loss |
|
| dgv11500n54 | Chr.7:90586963 - 90610884 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map