Assay Details
Target Gene Details
Entrez Gene ID: | 55536 |
Gene Name: | cell division cycle associated 7 like |
Gene Aliases: |
JPO2, R1, RAM2 |
Location: |
Chr.7:21900899-21945924 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDCA7L | NM_001127370.2 | NP_001120842.1 | ||
| NM_001127371.2 | NP_001120843.1 | |||
| NM_018719.4 | NP_061189.2 | |||
| AK022955.1 | BAB14330.1 | |||
| AK095965.1 | ||||
| AK292490.1 | ||||
| AK300781.1 | ||||
| AL359619.1 | CAB94887.1 | |||
| AY161168.1 | AAO17570.1 | |||
| AY161169.1 | AAO17571.1 | |||
| BC009352.2 | AAH09352.2 | |||
| BC014630.2 | AAH14630.2 | |||
| BC025242.1 | AAH25242.1 | |||
| BC032576.1 | AAH32576.1 | |||
| BC039823.1 | AAH39823.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830922 | Chr.7:21753768 - 21925057 on Build GRCh38 | Loss |
|
| nsv1027453 | Chr.7:21878856 - 21942830 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs150943620] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map