Assay Details
Target Gene Details
Entrez Gene ID: | 11112 |
Gene Name: | 3-hydroxyisobutyrate dehydrogenase |
Gene Aliases: |
NS5ATP1 |
Location: |
Chr.7:27525440-27663001 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIBADH | NM_152740.3 | NP_689953.1 | ||
| XM_017011719.1 | XP_016867208.1 | |||
| AB050000.1 | ||||
| AF529362.1 | AAQ09596.1 | |||
| AK025558.1 | ||||
| AK311650.1 | ||||
| AK316605.1 | ||||
| BC032324.1 | AAH32324.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs71539525] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map