Assay Details
Target Gene Details
Entrez Gene ID: | 4708 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit B2 |
Gene Aliases: |
AGGG, CI-AGGG |
Location: |
Chr.7:140696681-140706646 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFB2 | NM_004546.2 | NP_004537.1 | ||
| AF050639.1 | AAD05428.1 | |||
| AF067166.1 | AAD32450.1 | |||
| BC063026.1 | AAH63026.1 | |||
| CR542121.1 | CAG46918.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831158 | Chr.7:140692422 - 140902515 on Build GRCh38 | Loss |
|
| nsv831157 | Chr.7:140541539 - 140727076 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map