Assay Details
Target Gene Details
Entrez Gene ID: | 10551 |
Gene Name: | anterior gradient 2, protein disulphide isomerase family member |
Gene Aliases: |
AG2, GOB-4, HAG-2, HEL-S-116, PDIA17, XAG-2 |
Location: |
Chr.7:16791811-16805114 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGR2 | NM_006408.3 | NP_006399.1 | ||
| XM_005249581.4 | XP_005249638.1 | |||
| AF007791.1 | AAC77358.1 | |||
| AF038451.1 | AAC82614.1 | |||
| AF087879.1 | AAP97179.1 | |||
| AF088867.1 | AAF22484.1 | |||
| AF115926.1 | AAL54870.1 | |||
| AK223067.1 | BAD96787.1 | |||
| AK312942.1 | ||||
| AY359009.1 | AAQ89368.1 | |||
| BC015503.1 | AAH15503.1 | |||
| BF680655.1 | ||||
| BT007048.1 | AAP35697.1 | |||
| FJ224307.1 | ||||
| KJ767789.1 | ||||
| KJ767790.1 | ||||
| KJ767791.1 | ||||
| KJ767792.1 | ||||
| KJ767793.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891076 | Chr.7:16794737 - 17707004 on Build GRCh38 | Gain |
|
| dgv6289n100 | Chr.7:16230707 - 17452994 on Build GRCh38 | Gain |
|
| nsv1028125 | Chr.7:16489535 - 16807601 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map