Assay Details
Target Gene Details
Entrez Gene ID: | 57157 |
Gene Name: | putative homeodomain transcription factor 2 |
Gene Aliases: |
- |
Location: |
Chr.7:77798792-77957504 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHTF2 | NM_001127358.1 | NP_001120830.1 | ||
| NM_001127360.1 | NP_001120832.1 | |||
| XM_005250508.4 | XP_005250565.1 | |||
| XM_005250509.4 | XP_005250566.1 | |||
| XM_006716063.3 | XP_006716126.1 | |||
| XM_011516422.2 | XP_011514724.1 | |||
| XM_011516423.2 | XP_011514725.1 | |||
| XM_017012458.1 | XP_016867947.1 | |||
| XM_017012459.1 | XP_016867948.1 | |||
| AK090643.1 | ||||
| AK225854.1 | ||||
| AK308289.1 | ||||
| AL136883.1 | CAB66817.1 | |||
| BC067742.1 | AAH67742.1 | |||
| BP321440.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74487411] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map