Assay Details
Target Gene Details
Entrez Gene ID: | 84132 |
Gene Name: | ubiquitin specific peptidase 42 |
Gene Aliases: |
- |
Location: |
Chr.7:6078743-6161564 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP42 | NM_032172.2 | NP_115548.1 | ||
| XM_005249883.4 | XP_005249940.1 | |||
| XM_006715790.1 | XP_006715853.1 | |||
| XM_006715791.2 | XP_006715854.1 | |||
| XM_011515573.1 | XP_011513875.1 | |||
| XM_011515574.1 | XP_011513876.1 | |||
| XM_011515578.1 | XP_011513880.1 | |||
| AJ601395.1 | CAE53097.1 | |||
| AK022759.1 | BAB14232.1 | |||
| AK298164.1 | ||||
| AK309375.1 | ||||
| AY618868.1 | ||||
| BC060846.1 | AAH60846.2 | |||
| BC132862.1 | ||||
| DB457216.2 | ||||
| DR001777.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034916 | Chr.7:5895149 - 6119339 on Build GRCh38 | Gain |
|
| nsv436904 | Chr.7:5035574 - 6713282 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map