Assay Details
Target Gene Details
Entrez Gene ID: | 23382 |
Gene Name: | adenosylhomocysteinase like 2 |
Gene Aliases: |
ADOHCYASE3, IRBIT2 |
Location: |
Chr.7:129225014-129430211 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AHCYL2 | NM_001130720.2 | NP_001124192.1 | ||
| NM_015328.3 | NP_056143.1 | |||
| XM_017011906.1 | XP_016867395.1 | |||
| AB020635.1 | ||||
| BC008349.1 | AAH08349.1 | |||
| BC024325.1 | AAH24325.1 | |||
| BG700017.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525482 | Chr.7:129250971 - 129405350 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map