Assay Details
Target Gene Details
Entrez Gene ID: | 223117 |
Gene Name: | semaphorin 3D |
Gene Aliases: |
Sema-Z2, coll-2 |
Location: |
Chr.7:84995192-85187176 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEMA3D | NM_152754.2 | NP_689967.2 | ||
| XM_011515960.1 | XP_011514262.1 | |||
| XM_011515961.2 | XP_011514263.1 | |||
| XM_017011873.1 | XP_016867362.1 | |||
| AY358937.1 | AAQ89296.1 | |||
| BC029590.1 | AAH29590.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161578 | Chr.7:84946999 - 85038828 on Build GRCh38 | Deletion |
|
| esv2422324 | Chr.7:84900974 - 85315597 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map