Assay Details
Target Gene Details
Entrez Gene ID: | 26 |
Gene Name: | amine oxidase, copper containing 1 |
Gene Aliases: |
ABP, ABP1, DAO, DAO1, KAO |
Location: |
Chr.7:150824875-150861291 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AOC1 | XM_017011944.1 | XP_016867433.1 | ||
| XM_017011945.1 | XP_016867434.1 | |||
| BX648159.1 | 1 | 2212 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1030109 | Chr.7:150615828 - 151003922 on Build GRCh38 | Gain |
|
| esv2735390 | Chr.7:150829998 - 150842197 on Build GRCh38 | Deletion |
|
| nsv608990 | Chr.7:150264185 - 151010400 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map