Assay Details
Target Gene Details
Entrez Gene ID: | 9690 |
Gene Name: | ubiquitin protein ligase E3C |
Gene Aliases: |
HECTH2 |
Location: |
Chr.7:157138952-157269372 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE3C | NM_014671.2 | NP_055486.2 | ||
| XM_005249564.4 | XP_005249621.1 | |||
| XM_017012818.1 | XP_016868307.1 | |||
| AK295138.1 | ||||
| BC014029.2 | AAH14029.1 | |||
| BC026241.1 | AAH26241.1 | |||
| D13635.1 | ||||
| DB454915.1 |
More Information
Set Membership: |
|

Genomic Map