Assay Details
Target Gene Details
Entrez Gene ID: | 100134713 |
Gene Name: | NDUFB2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.7:140695336-140697077 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFB2-AS1 | NR_024454.1 | 2 | 1451 | |
| AK129584.1 | 2 | 1451 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831157 | Chr.7:140541539 - 140727076 on Build GRCh38 | Gain |
|
| nsv831158 | Chr.7:140692422 - 140902515 on Build GRCh38 | Loss |
|

Genomic Map