Assay Details
Target Gene Details
Entrez Gene ID: | 105375254 |
Gene Name: | uncharacterized LOC105375254 |
Gene Aliases: |
- |
Location: |
Chr.7:43025325-43114052 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105375254 | XM_017012919.1 | XP_016868408.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73690248] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map