Assay Details
Target Gene Details
Entrez Gene ID: | 105375396 |
Gene Name: | bax inhibitor 1-like |
Gene Aliases: |
- |
Location: |
Chr.7:92412550-92439562 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105375396 | NM_001317905.1 | NP_001304834.1 | ||
| XM_017011679.1 | XP_016867168.1 | |||
| KF595048.1 |

Genomic Map