Assay Details
Target Gene Details
Entrez Gene ID: | 4967 |
Gene Name: | oxoglutarate dehydrogenase |
Gene Aliases: |
AKGDH, E1k, OGDC |
Location: |
Chr.7:44606522-44709070 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OGDH | NM_001003941.2 | NP_001003941.1 | ||
| NM_001165036.1 | NP_001158508.1 | |||
| NM_002541.3 | NP_002532.2 | |||
| XM_005249759.4 | XP_005249816.1 | |||
| XM_005249761.3 | XP_005249818.1 | |||
| XM_011515408.2 | XP_011513710.1 | |||
| AK296400.1 | ||||
| AK304439.1 | ||||
| AK314179.1 | ||||
| AU140451.1 | ||||
| BC009580.1 | AAH09580.1 | |||
| D10523.1 | BAA01393.1 | |||
| DC401267.1 |
More Information
Set Membership: |
|

Genomic Map