Assay Details
Target Gene Details
Entrez Gene ID: | 8805 |
Gene Name: | tripartite motif containing 24 |
Gene Aliases: |
PTC6, RNF82, TF1A, TIF1, TIF1A, TIF1ALPHA, hTIF1 |
Location: |
Chr.7:138460334-138585588 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIM24 | NM_003852.3 | NP_003843.3 | ||
| NM_015905.2 | NP_056989.2 | |||
| AK075306.1 | ||||
| AK312610.1 | 1 | 124 | ||
| CB988362.1 | 1 | 100 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34048 | Chr.7:138039346 - 138528374 on Build GRCh38 | Loss |
|
| nsv831153 | Chr.7:138393434 - 138580903 on Build GRCh38 | Loss |
|
| esv2735212 | Chr.7:138060002 - 140415947 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs182495590] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map