Assay Details
Target Gene Details
Entrez Gene ID: | 10371 |
Gene Name: | semaphorin 3A |
Gene Aliases: |
COLL1, HH16, Hsema-I, Hsema-III, SEMA1, SEMAD, SEMAIII, SEMAL, SemD, coll-1 |
Location: |
Chr.7:83956846-84492724 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEMA3A | XM_005250110.3 | XP_005250167.1 | ||
| XM_005250111.4 | XP_005250168.1 | |||
| XM_006715839.3 | XP_006715902.1 | |||
| XM_011515734.2 | XP_011514036.1 | |||
| XM_017011673.1 | XP_016867162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1020784 | Chr.7:83948086 - 84057792 on Build GRCh38 | Loss |
|
| nsv607707 | Chr.7:83751716 - 84017500 on Build GRCh38 | Gain |
|
| esv3613949 | Chr.7:83822470 - 83987422 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map