Assay Details
Target Gene Details
Entrez Gene ID: | 84691 |
Gene Name: | family with sequence similarity 71 member F1 |
Gene Aliases: |
FAM137A, NYD-SP18 |
Location: |
Chr.7:128709061-128731743 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM71F1 | NM_001282788.1 | NP_001269717.1 | ||
| NM_001282789.1 | NP_001269718.1 | |||
| NM_032599.3 | NP_115988.1 | |||
| NR_104242.1 | ||||
| NR_104243.1 | ||||
| XM_017012743.1 | XP_016868232.1 | |||
| AF367470.1 | AAK53406.1 | |||
| AK093161.1 | BAC04080.1 | |||
| AK302221.1 | ||||
| AK310308.1 | ||||
| AK310426.1 | ||||
| AK313608.1 | ||||
| BC037248.1 | AAH37248.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2670312 | Chr.7:128499519 - 128999613 on Build GRCh38 | Deletion |
|
| esv2735137 | Chr.7:128499440 - 128999664 on Build GRCh38 | Deletion |
|

Genomic Map